Product Details

SNP ID
rs58959948
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.15:50557334 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCCCATCACAAACCAGGCAGCAAG[C/T]AAGGTATTGGGTGCTACTCAATCCA
Phenotype
MIM: 605692
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
TRPM7 PubMed Links

Gene Details

Gene
TRPM7
Gene Name
transient receptor potential cation channel subfamily M member 7
There are no transcripts associated with this gene.

View Full Product Details