Product Details

SNP ID
rs55897440
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:35329986 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAATCCCCATGTCTAGAGATTAATG[G/T]TGTAGATAGAGTTTACTGAAAGGTT
Phenotype
MIM: 107266
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CD22 PubMed Links

Gene Details

Gene
CD22
Gene Name
CD22 molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001185099.1 Intron NP_001172028.1
NM_001185100.1 Intron NP_001172029.1
NM_001185101.1 Intron NP_001172030.1
NM_001278417.1 Intron NP_001265346.1
NM_001771.3 Intron NP_001762.2

View Full Product Details