Product Details

SNP ID
rs67751448
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:73121336 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGGACTTTGCAGCCGAGAATCACC[A/C]TTCTCTTTCTTGTCTCCGAAACCGC
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
TMEM171 PubMed Links

Gene Details

Gene
TMEM171
Gene Name
transmembrane protein 171
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001161342.2 Intron NP_001154814.1
NM_173490.7 Intron NP_775761.4
XM_011543156.1 Intron XP_011541458.1

View Full Product Details