Product Details
- SNP ID
-
rs73705088
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:120541149 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCAAGTGAAGTCATATACTTTTTCA[C/T]TATGGCAGGCAGAAACCATCTCAGG
- Phenotype
-
MIM: 600026
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
SNTB1
PubMed Links
Gene Details
- Gene
- SNTB1
- Gene Name
- syntrophin beta 1
View Full Product Details