Product Details

SNP ID
rs73299946
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:90994540 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAACATTTGTGATTAAGCAATTTCT[G/T]TTTCAAAACTGCAAAGATGGGCAGT
Phenotype
MIM: 609864
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
C8orf88 PubMed Links

Gene Details

Gene
C8orf88
Gene Name
chromosome 8 open reading frame 88
There are no transcripts associated with this gene.

Gene
TMEM55A
Gene Name
transmembrane protein 55A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018710.2 2137 UTR 3 NP_061180.1
XM_011517152.2 2137 Intron XP_011515454.1

View Full Product Details