Product Details

SNP ID
rs73480998
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:33702198 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTGCGATAGCTCAGCACCCAGCCT[A/G]TAATCCCAGCACTCTGGGAGTCTGA
Phenotype
MIM: 107271
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C11orf91 PubMed Links

Gene Details

Gene
C11orf91
Gene Name
chromosome 11 open reading frame 91
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001166692.1 Intron NP_001160164.1
XM_017017053.1 Intron XP_016872542.1
Gene
CD59
Gene Name
CD59 molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000611.5 Intron NP_000602.1
NM_001127223.1 Intron NP_001120695.1
NM_001127225.1 Intron NP_001120697.1
NM_001127226.1 Intron NP_001120698.1
NM_001127227.1 Intron NP_001120699.1
NM_203329.2 Intron NP_976074.1
NM_203330.2 Intron NP_976075.1
NM_203331.2 Intron NP_976076.1

View Full Product Details