Product Details

SNP ID
rs72773107
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:56996035 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGAAAGCCTCAGTCAGGTGCTAAC[A/G]CATCTGTAAACCTCTCACACTCTTG
Phenotype
MIM: 613537
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NLRC5 PubMed Links

Gene Details

Gene
NLRC5
Gene Name
NLR family CARD domain containing 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032206.4 Intron NP_115582.4
XM_005256193.2 Intron XP_005256250.1
XM_005256194.3 Intron XP_005256251.1
XM_005256195.2 Intron XP_005256252.1
XM_005256196.2 Intron XP_005256253.1
XM_005256197.2 Intron XP_005256254.1
XM_005256199.3 Intron XP_005256256.1
XM_005256201.3 Intron XP_005256258.1
XM_006721297.2 Intron XP_006721360.1
XM_006721298.2 Intron XP_006721361.1
XM_006721300.2 Intron XP_006721363.1
XM_011523373.2 Intron XP_011521675.1
XM_011523375.2 Intron XP_011521677.1
XM_011523376.2 Intron XP_011521678.1
XM_017023766.1 Intron XP_016879255.1
XM_017023767.1 Intron XP_016879256.1
XM_017023768.1 Intron XP_016879257.1
XM_017023769.1 Intron XP_016879258.1
XM_017023770.1 Intron XP_016879259.1
XM_017023771.1 Intron XP_016879260.1

View Full Product Details