Product Details
- SNP ID
-
rs73351760
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:67826204 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCGAGGAGACCCAGGATTCTGAGGA[C/G]GACGAGGAGGATGAGATGGAAGAGG
- Phenotype
-
MIM: 601819
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
BPTF
PubMed Links
Gene Details
- Gene
- BPTF
- Gene Name
- bromodomain PHD finger transcription factor
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_004459.6 |
688 |
Missense Mutation |
GAC,GAG |
D160E |
NP_004450.3 |
NM_182641.3 |
688 |
Missense Mutation |
GAC,GAG |
D160E |
NP_872579.2 |
XM_005257150.3 |
688 |
Missense Mutation |
GAC,GAG |
D160E |
XP_005257207.1 |
XM_005257151.3 |
688 |
Missense Mutation |
GAC,GAG |
D160E |
XP_005257208.1 |
XM_005257152.2 |
688 |
Missense Mutation |
GAC,GAG |
D160E |
XP_005257209.1 |
XM_005257153.3 |
688 |
Missense Mutation |
GAC,GAG |
D160E |
XP_005257210.1 |
XM_005257154.3 |
688 |
Missense Mutation |
GAC,GAG |
D160E |
XP_005257211.1 |
XM_005257155.3 |
688 |
Missense Mutation |
GAC,GAG |
D160E |
XP_005257212.1 |
XM_005257156.3 |
688 |
Missense Mutation |
GAC,GAG |
D160E |
XP_005257213.1 |
XM_005257157.3 |
688 |
Missense Mutation |
GAC,GAG |
D160E |
XP_005257214.1 |
XM_005257158.3 |
688 |
Missense Mutation |
GAC,GAG |
D160E |
XP_005257215.1 |
XM_005257159.2 |
688 |
Missense Mutation |
GAC,GAG |
D160E |
XP_005257216.1 |
XM_005257160.2 |
688 |
Missense Mutation |
GAC,GAG |
D160E |
XP_005257217.1 |
XM_005257161.3 |
688 |
Missense Mutation |
GAC,GAG |
D160E |
XP_005257218.1 |
XM_011524520.2 |
688 |
Missense Mutation |
GAC,GAG |
D160E |
XP_011522822.1 |
XM_011524521.2 |
688 |
Missense Mutation |
GAC,GAG |
D160E |
XP_011522823.1 |
XM_011524522.2 |
688 |
Missense Mutation |
GAC,GAG |
D160E |
XP_011522824.1 |
XM_011524523.2 |
688 |
Missense Mutation |
GAC,GAG |
D160E |
XP_011522825.1 |
XM_011524524.2 |
688 |
Intron |
|
|
XP_011522826.1 |
XM_011524525.2 |
688 |
Missense Mutation |
GAC,GAG |
D160E |
XP_011522827.1 |
XM_011524526.2 |
688 |
Missense Mutation |
GAC,GAG |
D160E |
XP_011522828.1 |
XM_017024353.1 |
688 |
Missense Mutation |
GAC,GAG |
D160E |
XP_016879842.1 |
XM_017024354.1 |
688 |
Intron |
|
|
XP_016879843.1 |
View Full Product Details