Product Details

SNP ID
rs73563879
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:6381134 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCTGGGCCTTACCGCCATGAAGAG[A/G]GCTGAGGAGGCCTCGCTGTCAATGT
Phenotype
MIM: 613305 MIM: 189968 MIM: 602921
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ALKBH7 PubMed Links

Gene Details

Gene
ALKBH7
Gene Name
alkB homolog 7
There are no transcripts associated with this gene.

Gene
GTF2F1
Gene Name
general transcription factor IIF subunit 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002096.2 1365 Silent Mutation GCC,GCT A360A NP_002087.2
Gene
MIR6885
Gene Name
microRNA 6885
There are no transcripts associated with this gene.

Gene
PSPN
Gene Name
persephin
There are no transcripts associated with this gene.

View Full Product Details