Product Details

SNP ID
rs71647827
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.20:3889381 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAAGAGGCGGCCGGCCGAGGGCGC[C/G]CCTCTGCTCTGGCTGGACTGCCGCG
Phenotype
MIM: 606157
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
PANK2 PubMed Links
Additional Information
For this assay, SNP(s) [rs71647828] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
PANK2
Gene Name
pantothenate kinase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001324191.1 448 UTR 5 NP_001311120.1
NM_001324192.1 448 Missense Mutation CCC,CGC P94R NP_001311121.1
NM_001324193.1 448 Intron NP_001311122.1
NM_024960.5 448 Intron NP_079236.3
NM_153638.3 448 Missense Mutation CCC,CGC P94R NP_705902.2
NM_153640.3 448 Intron NP_705904.1
XM_005260835.3 448 Intron XP_005260892.1
XM_005260836.4 448 Intron XP_005260893.3
XM_011529364.2 448 Missense Mutation CCC,CGC P94R XP_011527666.1
XM_011529365.2 448 Missense Mutation CCC,CGC P94R XP_011527667.1
XM_017028077.1 448 Intron XP_016883566.1
XM_017028078.1 448 Intron XP_016883567.1
XM_017028079.1 448 Intron XP_016883568.1

View Full Product Details