Product Details
- SNP ID
-
rs74148798
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
13
- Location
-
Chr.1:206802873 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAATCTTCTGGCAGATAAGAAGGAC[G/T]GTTAAGGAAGACGAGGGGACAGGAA
- Phenotype
-
MIM: 605687
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
IL19
PubMed Links
Gene Details
- Gene
- IL19
- Gene Name
- interleukin 19
View Full Product Details