Product Details
- SNP ID
-
rs73551679
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:107503598 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTAACCTAAGTGGTAAGTTAACTGG[G/T]TTACTATTATGCAATATTGTACAGA
- Phenotype
-
MIM: 613306
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
ALKBH8
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs560973772] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ALKBH8
- Gene Name
- alkB homolog 8, tRNA methyltransferase
View Full Product Details