Product Details

SNP ID
rs3918367
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.3:46357683 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTGCCTCCGCTCTACTCGCTGGT[G/T]TTCATCTTTGGTTTTGTGGGCAACA
Phenotype
MIM: 601267
Polymorphism
G/T, Transversion Substitution
Allele Nomenclature
Literature Links
CCR2 PubMed Links
Additional Information
For this assay, SNP(s) [rs1799864] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
CCR2
Gene Name
C-C motif chemokine receptor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001123041.2 354 Silent Mutation GTG,GTT V52V NP_001116513.2
NM_001123396.1 354 Silent Mutation GTG,GTT V52V NP_001116868.1
XM_011534069.1 354 Silent Mutation GTG,GTT V52V XP_011532371.1
Gene
LOC102724297
Gene Name
uncharacterized LOC102724297
There are no transcripts associated with this gene.

View Full Product Details