Product Details

SNP ID
rs77755255
Assay Type
Functionally tested
NCBI dbSNP Submissions
13
Location
Chr.1:26863705 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATCAGCAAGAAGGAGATGCCGCCC[A/T]CCAACCCCATCCGCCTGGGCCTGGC
Phenotype
MIM: 601290
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
SFN PubMed Links

Gene Details

Gene
SFN
Gene Name
stratifin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006142.3 564 Missense Mutation ACC,TCC T165S NP_006133.1
Gene
ZDHHC18
Gene Name
zinc finger DHHC-type containing 18
There are no transcripts associated with this gene.

View Full Product Details