Product Details

SNP ID
rs251248
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:173060115 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTAAGTTTGTGTGTTGATTCTGTTA[A/G]TATAACTCAGTTTTTAAAAGATAAA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CREBRF PubMed Links

Gene Details

Gene
CREBRF
Gene Name
CREB3 regulatory factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001168393.1 Intron NP_001161865.1
NM_001168394.1 Intron NP_001161866.1
NM_153607.2 Intron NP_705835.2
XM_005265821.3 Intron XP_005265878.1
XM_006714822.3 Intron XP_006714885.1
XM_011534442.1 Intron XP_011532744.1

View Full Product Details