Product Details

SNP ID
rs532244
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:104947734 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGTGATAAAAATCTATTGTACAGG[A/G]TGGTGCCAGTAGTAACCAACATTGT
Phenotype
MIM: 602664
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CASP4 PubMed Links

Gene Details

Gene
CASP4
Gene Name
caspase 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001225.3 Intron NP_001216.1
NM_033306.2 Intron NP_150649.1
XM_011543019.1 Intron XP_011541321.1
XM_017018397.1 Intron XP_016873886.1

View Full Product Details