Product Details
- SNP ID
-
rs1190274
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:105147386 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TGAGGACAACACTTTTAAAGGTACT[A/G]TTTTATTTAGTGTGTAAACAACTTC
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
BVES-AS1
PubMed Links
Gene Details
- Gene
- BVES-AS1
- Gene Name
- BVES antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details