Product Details
- SNP ID
-
rs1566879
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:10587352 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AGAAAAAGATGCCTCTTCTGAGCCA[A/G]TGATTTAGAAAATAGTAACCTCTCA
- Phenotype
-
MIM: 602334
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
EMP2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs116251542] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- EMP2
- Gene Name
- epithelial membrane protein 2
There are no transcripts associated with this gene.
- Gene
- TEKT5
- Gene Name
- tektin 5
View Full Product Details