Product Details

SNP ID
rs4908540
Assay Type
Functionally tested
NCBI dbSNP Submissions
32
Location
Chr.1:6206036 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCTGGGGAAGGTGGATGCCGGAAG[A/G]GGCGTCGCCTGCGGGTCACCCAGAG
Phenotype
MIM: 616923 MIM: 180474
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC102724450 PubMed Links

Gene Details

Gene
LOC102724450
Gene Name
uncharacterized LOC102724450
There are no transcripts associated with this gene.

Gene
RNF207
Gene Name
ring finger protein 207
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_207396.2 562 Intron NP_997279.2
XM_011541439.2 562 Missense Mutation GAG,GGG E4G XP_011539741.1
XM_017001259.1 562 Intron XP_016856748.1
Gene
RPL22
Gene Name
ribosomal protein L22
There are no transcripts associated with this gene.

View Full Product Details