Product Details

SNP ID
rs3744372
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:35475137 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCTTGTCTTGCCTTCAGGGCTCAAG[G/T]AGAAGATCTTTGTCATGACACACAC
Phenotype
MIM: 614956
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
SLFN12L PubMed Links

Gene Details

Gene
SLFN12L
Gene Name
schlafen family member 12 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001195790.1 8304 Missense Mutation TAC,TCC Y518S NP_001182719.1
XM_017024005.1 8304 Missense Mutation TAC,TCC Y547S XP_016879494.1
XM_017024006.1 8304 Missense Mutation TAC,TCC Y518S XP_016879495.1
XM_017024007.1 8304 Missense Mutation TAC,TCC Y518S XP_016879496.1
XM_017024008.1 8304 Missense Mutation TAC,TCC Y500S XP_016879497.1
XM_017024009.1 8304 Missense Mutation TAC,TCC Y500S XP_016879498.1

View Full Product Details