Product Details

SNP ID
rs11793162
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:129806893 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAGTGAGACCCTGTTTCCAAAGACA[A/G]AAAAGAGAAGAAAAGAATCACGGAT
Phenotype
MIM: 605204 MIM: 608050
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TOR1A PubMed Links

Gene Details

Gene
TOR1A
Gene Name
torsin family 1 member A
There are no transcripts associated with this gene.

Gene
TOR1B
Gene Name
torsin family 1 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317893.1 Intron NP_001304822.1
NM_001317894.1 Intron NP_001304823.1
NM_014506.2 Intron NP_055321.1
XM_005251927.3 Intron XP_005251984.1

View Full Product Details