Product Details
- SNP ID
-
rs8150
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:76470935 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- TTATTTAATCCATGAGACATCGTCC[C/G]TACAAAGTTAGCGTTACTTTTAAAA
- Phenotype
-
MIM: 600950
MIM: 614404
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
AANAT
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs7221225] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- AANAT
- Gene Name
- aralkylamine N-acetyltransferase
- Gene
- RHBDF2
- Gene Name
- rhomboid 5 homolog 2
View Full Product Details