Product Details

SNP ID
rs31519
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:135950748 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTGAGTAAATGCAAGTATCTATCC[G/A]TTATGGTTAGCATTGCTGAAAGTGG
Phenotype
MIM: 609087 MIM: 602882
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
FBXL21 PubMed Links

Gene Details

Gene
FBXL21
Gene Name
F-box and leucine rich repeat protein 21 (gene/pseudogene)
There are no transcripts associated with this gene.

Gene
LECT2
Gene Name
leukocyte cell derived chemotaxin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002302.2 Intron NP_002293.2

View Full Product Details