Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_002817.3 | 137 | UTR 5 | NP_002808.3 | ||
NM_175932.2 | 137 | UTR 5 | NP_787128.2 | ||
XM_011520235.2 | 137 | UTR 5 | XP_011518537.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001017524.2 | 137 | Intron | NP_001017524.1 | ||
NM_012239.5 | 137 | Intron | NP_036371.1 | ||
XM_005252835.1 | 137 | Intron | XP_005252892.1 | ||
XM_011519956.1 | 137 | Intron | XP_011518258.1 | ||
XM_011519957.1 | 137 | Intron | XP_011518259.1 | ||
XM_017017428.1 | 137 | Intron | XP_016872917.1 | ||
XM_017017429.1 | 137 | UTR 5 | XP_016872918.1 | ||
XM_017017430.1 | 137 | UTR 5 | XP_016872919.1 | ||
XM_017017431.1 | 137 | Intron | XP_016872920.1 |