Product Details

SNP ID
rs363179
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:66757661 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTTGAAAGTTTTGGCAGTGAAAA[C/T]AGCCAAATAAATATATGTGGCAAAA
Phenotype
MIM: 611406
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DYNC1LI2 PubMed Links

Gene Details

Gene
DYNC1LI2
Gene Name
dynein cytoplasmic 1 light intermediate chain 2
There are no transcripts associated with this gene.

Gene
LOC106699570
Gene Name
uncharacterized LOC106699570
There are no transcripts associated with this gene.

Gene
TERB1
Gene Name
telomere repeat binding bouquet formation protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136505.1 Intron NP_001129977.1
XM_011523004.2 Intron XP_011521306.1
XM_011523005.2 Intron XP_011521307.1
XM_011523006.2 Intron XP_011521308.1
XM_011523007.2 Intron XP_011521309.1
XM_011523008.2 Intron XP_011521310.1
XM_011523009.2 Intron XP_011521311.1
XM_011523010.2 Intron XP_011521312.1
XM_011523011.2 Intron XP_011521313.1
XM_011523012.2 Intron XP_011521314.1
XM_011523014.2 Intron XP_011521316.1
XM_011523015.2 Intron XP_011521317.1
XM_011523016.2 Intron XP_011521318.1
XM_011523017.1 Intron XP_011521319.1
XM_011523018.1 Intron XP_011521320.1
XM_011523020.1 Intron XP_011521322.1
XM_017023157.1 Intron XP_016878646.1
XM_017023158.1 Intron XP_016878647.1
XM_017023159.1 Intron XP_016878648.1

View Full Product Details