Product Details
- SNP ID
-
rs363181
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:66759249 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TATGGGCAAGAGTGCAAGAGCTTGC[A/T]AAAGTTTCGGCTATTCAGGGATTTT
- Phenotype
-
MIM: 611406
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
DYNC1LI2
PubMed Links
Gene Details
- Gene
- DYNC1LI2
- Gene Name
- dynein cytoplasmic 1 light intermediate chain 2
There are no transcripts associated with this gene.
- Gene
- LOC106699570
- Gene Name
- uncharacterized LOC106699570
There are no transcripts associated with this gene.
- Gene
- TERB1
- Gene Name
- telomere repeat binding bouquet formation protein 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001136505.1 |
2084 |
Missense Mutation |
AGC,TGC |
S608C |
NP_001129977.1 |
XM_011523004.2 |
2084 |
Missense Mutation |
AGC,TGC |
S608C |
XP_011521306.1 |
XM_011523005.2 |
2084 |
Missense Mutation |
AGC,TGC |
S608C |
XP_011521307.1 |
XM_011523006.2 |
2084 |
Missense Mutation |
AGC,TGC |
S608C |
XP_011521308.1 |
XM_011523007.2 |
2084 |
Missense Mutation |
AGC,TGC |
S608C |
XP_011521309.1 |
XM_011523008.2 |
2084 |
Missense Mutation |
AGC,TGC |
S608C |
XP_011521310.1 |
XM_011523009.2 |
2084 |
Missense Mutation |
AGC,TGC |
S608C |
XP_011521311.1 |
XM_011523010.2 |
2084 |
Missense Mutation |
AGC,TGC |
S597C |
XP_011521312.1 |
XM_011523011.2 |
2084 |
Missense Mutation |
AGC,TGC |
S597C |
XP_011521313.1 |
XM_011523012.2 |
2084 |
Missense Mutation |
AGC,TGC |
S597C |
XP_011521314.1 |
XM_011523014.2 |
2084 |
Intron |
|
|
XP_011521316.1 |
XM_011523015.2 |
2084 |
Missense Mutation |
AGC,TGC |
S608C |
XP_011521317.1 |
XM_011523016.2 |
2084 |
Missense Mutation |
AGC,TGC |
S608C |
XP_011521318.1 |
XM_011523017.1 |
2084 |
Missense Mutation |
AGC,TGC |
S608C |
XP_011521319.1 |
XM_011523018.1 |
2084 |
Intron |
|
|
XP_011521320.1 |
XM_011523020.1 |
2084 |
Intron |
|
|
XP_011521322.1 |
XM_017023157.1 |
2084 |
Intron |
|
|
XP_016878646.1 |
XM_017023158.1 |
2084 |
Intron |
|
|
XP_016878647.1 |
XM_017023159.1 |
2084 |
Intron |
|
|
XP_016878648.1 |
View Full Product Details