Product Details

SNP ID
rs2236737
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.17:63860740 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
AAGAATCATACCCATTGGCAGGGAC[C/T]GACATTAATGTGACCTGCTCAGGGC
Phenotype
MIM: 612756
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TCAM1P PubMed Links

Gene Details

Gene
TCAM1P
Gene Name
testicular cell adhesion molecule 1, pseudogene
There are no transcripts associated with this gene.

View Full Product Details