Product Details
- SNP ID
-
rs10404537
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:1289553 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CATCAAGCCTCAGAGCAGGGTGGAC[A/G]CAGGGGCCTGTGGTCCCCGGGAGTC
- Phenotype
-
MIM: 602756
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
EFNA2
PubMed Links
Gene Details
- Gene
- EFNA2
- Gene Name
- ephrin A2
View Full Product Details