Product Details
- SNP ID
-
rs11881588
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:1291871 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAGGAGGGGAGCCCTGCTGGGTGAG[C/G]CTCTGCCTGGGGCCTGGAGGGCTGC
- Phenotype
-
MIM: 602756
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
EFNA2
PubMed Links
Gene Details
- Gene
- EFNA2
- Gene Name
- ephrin A2
View Full Product Details