Product Details

SNP ID
rs1799865
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.3:46358307 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
ACTTTCTCTTCTGGACTCCCTATAA[C/T]ATTGTCATTCTCCTGAACACCTTCC
Phenotype
MIM: 601267
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCR2 PubMed Links

Gene Details

Gene
CCR2
Gene Name
C-C motif chemokine receptor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001123041.2 978 Silent Mutation AAC,AAT N260N NP_001116513.2
NM_001123396.1 978 Silent Mutation AAC,AAT N260N NP_001116868.1
XM_011534069.1 978 Silent Mutation AAC,AAT N260N XP_011532371.1
Gene
LOC102724297
Gene Name
uncharacterized LOC102724297
There are no transcripts associated with this gene.

View Full Product Details