Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001317237.1 | 2113 | UTR 3 | NP_001304166.1 | ||
NM_001908.4 | 2113 | UTR 3 | NP_001899.1 | ||
NM_147780.3 | 2113 | UTR 3 | NP_680090.1 | ||
NM_147781.3 | 2113 | UTR 3 | NP_680091.1 | ||
NM_147782.3 | 2113 | UTR 3 | NP_680092.1 | ||
NM_147783.3 | 2113 | UTR 3 | NP_680093.1 | ||
XM_006716244.2 | 2113 | UTR 3 | XP_006716307.1 | ||
XM_006716245.2 | 2113 | UTR 3 | XP_006716308.1 | ||
XM_011543812.2 | 2113 | UTR 3 | XP_011542114.1 | ||
XM_017013097.1 | 2113 | UTR 3 | XP_016868586.1 | ||
XM_017013098.1 | 2113 | UTR 3 | XP_016868587.1 | ||
XM_017013099.1 | 2113 | UTR 3 | XP_016868588.1 | ||
XM_017013100.1 | 2113 | UTR 3 | XP_016868589.1 | ||
XM_017013101.1 | 2113 | UTR 3 | XP_016868590.1 |