Product Details
- SNP ID
-
rs1053561
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:85318929 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TATGACTGAAGCACAGATGTGTCTA[A/G]TAGAAATCACCCTTCACCCAAAAGC
- Phenotype
-
MIM: 604652
MIM: 603062
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
TCF7L1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs116012131] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- TCF7L1
- Gene Name
- transcription factor 7 like 1
There are no transcripts associated with this gene.
- Gene
- TGOLN2
- Gene Name
- trans-golgi network protein 2
View Full Product Details