Product Details

SNP ID
rs10077757
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.5:95854901 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
TACTGTTTGTTGACTGTTCACCAAG[A/T]ATCAGGCACTCGGCTAAGAATTGTT
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
LINC01554 PubMed Links

Gene Details

Gene
LINC01554
Gene Name
long intergenic non-protein coding RNA 1554
There are no transcripts associated with this gene.

Gene
LOC102724720
Gene Name
uncharacterized LOC102724720
There are no transcripts associated with this gene.

View Full Product Details