Product Details

SNP ID
rs71365437
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.19:54218979 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTATGAGATAGAAAAAAACTCCC[A/G]TGAATACTGAAGTTTGTAAATGCGT
Phenotype
MIM: 604820
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
LILRB3 PubMed Links
Additional Information
For this assay, SNP(s) [rs117563200] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
LILRB3
Gene Name
leukocyte immunoglobulin like receptor B3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001081450.2 Intron NP_001074919.2
NM_001320960.1 Intron NP_001307889.1
NM_006864.3 Intron NP_006855.3
XM_011526381.2 Intron XP_011524683.1
XM_011526382.2 Intron XP_011524684.1
XM_017026218.1 Intron XP_016881707.1
XM_017026219.1 Intron XP_016881708.1
XM_017026220.1 Intron XP_016881709.1
XM_017026221.1 Intron XP_016881710.1
XM_017026222.1 Intron XP_016881711.1
XM_017026223.1 Intron XP_016881712.1

View Full Product Details