Product Details

SNP ID
rs7205459
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.16:56991259 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TGCTGAACTTTATTCCTGGATCAGA[C/T]ACAGAGCTGTCCGGGTGTCCACAGG
Phenotype
MIM: 118470 MIM: 613537
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CETP PubMed Links

Gene Details

Gene
CETP
Gene Name
cholesteryl ester transfer protein
There are no transcripts associated with this gene.

Gene
NLRC5
Gene Name
NLR family CARD domain containing 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032206.4 Intron NP_115582.4
XM_005256193.2 Intron XP_005256250.1
XM_005256194.3 Intron XP_005256251.1
XM_005256195.2 Intron XP_005256252.1
XM_005256196.2 Intron XP_005256253.1
XM_005256197.2 Intron XP_005256254.1
XM_005256199.3 Intron XP_005256256.1
XM_005256201.3 Intron XP_005256258.1
XM_006721297.2 Intron XP_006721360.1
XM_006721298.2 Intron XP_006721361.1
XM_006721300.2 Intron XP_006721363.1
XM_011523373.2 Intron XP_011521675.1
XM_011523375.2 Intron XP_011521677.1
XM_011523376.2 Intron XP_011521678.1
XM_017023766.1 Intron XP_016879255.1
XM_017023767.1 Intron XP_016879256.1
XM_017023768.1 Intron XP_016879257.1
XM_017023769.1 Intron XP_016879258.1
XM_017023770.1 Intron XP_016879259.1
XM_017023771.1 Intron XP_016879260.1

View Full Product Details