Product Details

SNP ID
rs3758337
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:129835822 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAAGCATCCGCGTGGCGTCTCGCGT[C/T]TCGGGCGCTGACTTTTCTTTCCTCG
Phenotype
MIM: 615143
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C9orf78 PubMed Links

Gene Details

Gene
C9orf78
Gene Name
chromosome 9 open reading frame 78
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016520.2 336 Intron NP_057604.1
XM_011518780.2 336 Intron XP_011517082.1
Gene
USP20
Gene Name
ubiquitin specific peptidase 20
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001008563.4 336 Intron NP_001008563.2
NM_001110303.3 336 Intron NP_001103773.2
NM_006676.7 336 Intron NP_006667.3
XM_005251665.3 336 Intron XP_005251722.1
XM_005251666.3 336 Intron XP_005251723.1
XM_011518161.2 336 Intron XP_011516463.1
XM_011518162.2 336 UTR 5 XP_011516464.1
XM_017014221.1 336 Intron XP_016869710.1

View Full Product Details