Product Details

SNP ID
rs6931809
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:24706542 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTCATGGGGACAGATTTTGAGAAGT[C/T]TGAAGTCATTTTTATATAGAGGAAG
Phenotype
MIM: 615652
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ACOT13 PubMed Links

Gene Details

Gene
ACOT13
Gene Name
acyl-CoA thioesterase 13
There are no transcripts associated with this gene.

Gene
C6orf62
Gene Name
chromosome 6 open reading frame 62
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030939.4 Intron NP_112201.1
XM_005249433.3 Intron XP_005249490.1
XM_011514928.2 Intron XP_011513230.1

View Full Product Details