Product Details

SNP ID
rs1000245
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:18920278 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTGCTCCTCTAGCCCAACCCAGTTT[C/G]GCCATTAAAGAGAGGCCCACAGGAA
Phenotype
MIM: 606942 MIM: 604800
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
COPE PubMed Links

Gene Details

Gene
COPE
Gene Name
coatomer protein complex subunit epsilon
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007263.3 Intron NP_009194.2
NM_199442.1 Intron NP_955474.1
NM_199444.1 Intron NP_955476.1
XM_011527655.1 Intron XP_011525957.1
XM_011527656.1 Intron XP_011525958.1
Gene
DDX49
Gene Name
DEAD-box helicase 49
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019070.4 Intron NP_061943.2
XM_011528083.1 Intron XP_011526385.1
XM_011528084.2 Intron XP_011526386.1
Gene
HOMER3
Gene Name
homer scaffolding protein 3
There are no transcripts associated with this gene.

View Full Product Details