Product Details

SNP ID
rs853575
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:118059844 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAGTTACCCAAGAAGATCCCTTTTC[C/T]CTGGAAGTGTTAGGTCTCCAAAGGA
Phenotype
MIM: 608598
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CASC2 PubMed Links

Gene Details

Gene
CASC2
Gene Name
cancer susceptibility candidate 2 (non-protein coding)
There are no transcripts associated with this gene.

View Full Product Details