Product Details
- SNP ID
-
rs853565
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:118054008 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACTTTATGCACGGACAAAGGGTGAC[A/C]TAGGATATCACTGAACTCTTCTGCC
- Phenotype
-
MIM: 608598
MIM: 608599
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
CASC2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs78679797] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CASC2
- Gene Name
- cancer susceptibility candidate 2 (non-protein coding)
There are no transcripts associated with this gene.
- Gene
- RAB11FIP2
- Gene Name
- RAB11 family interacting protein 2
There are no transcripts associated with this gene.
View Full Product Details