Product Details

SNP ID
rs1619657
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:2482438 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGGGAAGTGTCGAGGATGTTGAC[C/G]GCCTCTAGCATCTGAGGTACATCTG
Phenotype
MIM: 602850
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
RNF4 PubMed Links

Gene Details

Gene
RNF4
Gene Name
ring finger protein 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001185009.2 Intron NP_001171938.1
NM_001185010.2 Intron NP_001171939.1
NM_002938.4 Intron NP_002929.1

View Full Product Details