Product Details

SNP ID
rs1062721
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:55510694 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCTTCAAACAACAGATTAAAGAAA[A/T]AGAGGAGGGTAAAGGCTCGCAATTT
Phenotype
MIM: 164975
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ERC2 PubMed Links

Gene Details

Gene
ERC2
Gene Name
ELKS/RAB6-interacting/CAST family member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015576.2 3816 UTR 3 NP_056391.1
XM_017006138.1 3816 UTR 3 XP_016861627.1
XM_017006139.1 3816 UTR 3 XP_016861628.1
XM_017006140.1 3816 UTR 3 XP_016861629.1
XM_017006141.1 3816 UTR 3 XP_016861630.1
XM_017006142.1 3816 UTR 3 XP_016861631.1
XM_017006143.1 3816 UTR 3 XP_016861632.1
XM_017006144.1 3816 UTR 3 XP_016861633.1
XM_017006145.1 3816 UTR 3 XP_016861634.1
XM_017006146.1 3816 UTR 3 XP_016861635.1
XM_017006147.1 3816 UTR 3 XP_016861636.1
XM_017006148.1 3816 UTR 3 XP_016861637.1
XM_017006149.1 3816 UTR 3 XP_016861638.1
XM_017006150.1 3816 UTR 3 XP_016861639.1
XM_017006151.1 3816 UTR 3 XP_016861640.1
XM_017006152.1 3816 UTR 3 XP_016861641.1
XM_017006153.1 3816 UTR 3 XP_016861642.1
XM_017006154.1 3816 UTR 3 XP_016861643.1
XM_017006155.1 3816 UTR 3 XP_016861644.1
XM_017006156.1 3816 UTR 3 XP_016861645.1
XM_017006157.1 3816 UTR 3 XP_016861646.1
XM_017006158.1 3816 UTR 3 XP_016861647.1
XM_017006159.1 3816 UTR 3 XP_016861648.1
Gene
WNT5A
Gene Name
Wnt family member 5A
There are no transcripts associated with this gene.

View Full Product Details