Product Details

SNP ID
rs866477
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:74265553 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGACTTTTACTTTGCAGAACTTAG[C/T]TCTGCAATAAGTAGTTCCTGTTTTC
Phenotype
MIM: 601325
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CNTN3 PubMed Links

Gene Details

Gene
CNTN3
Gene Name
contactin 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020872.2 Intron NP_065923.1
XM_005264757.3 Intron XP_005264814.1
XM_011533768.2 Intron XP_011532070.1
XM_017006507.1 Intron XP_016861996.1
XM_017006508.1 Intron XP_016861997.1
XM_017006509.1 Intron XP_016861998.1

View Full Product Details