Product Details

SNP ID
rs1373199
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.2:68368548 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TGCAAAAGAACTCACTAGAGCCAGA[A/G]CAAGGCACGAACAAATTGTTTTGTG
Phenotype
MIM: 173570
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC101927723 PubMed Links

Gene Details

Gene
LOC101927723
Gene Name
uncharacterized LOC101927723
There are no transcripts associated with this gene.

Gene
PLEK
Gene Name
pleckstrin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002664.2 Intron NP_002655.2

View Full Product Details