Product Details
- SNP ID
-
rs289752
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:56993024 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CACCCAATTGCCCTCCCCGAAGACA[A/G]CCAATGTTATCAGGTTCTTGTGTCT
- Phenotype
-
MIM: 118470
MIM: 613537
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CETP
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs77431602] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CETP
- Gene Name
- cholesteryl ester transfer protein
There are no transcripts associated with this gene.
- Gene
- NLRC5
- Gene Name
- NLR family CARD domain containing 5
View Full Product Details