Product Details
- SNP ID
-
rs1533849
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
40
- Location
-
Chr.1:76881279 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTTCAAATTCTTCTCCATAAGCTCC[A/G]AGTCTTAAACACTTAGTCTGGGCCT
- Phenotype
-
MIM: 610134
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ST6GALNAC5
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs76615695,rs79035511] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ST6GALNAC5
- Gene Name
- ST6 N-acetylgalactosaminide alpha-2,6-sialyltransferase 5
View Full Product Details