Product Details

SNP ID
rs2280601
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:99458476 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAGAATTCCCTTCAGAGATGACCCA[C/T]GACCCGCCTGCCGGTGTCTCTGCAG
Phenotype
MIM: 603052
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ATP5J2 PubMed Links

Gene Details

Gene
ATP5J2
Gene Name
ATP synthase, H+ transporting, mitochondrial Fo complex subunit F2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001003713.2 Intron NP_001003713.1
NM_001003714.2 Intron NP_001003714.1
NM_001039178.2 Intron NP_001034267.1
NM_004889.3 Intron NP_004880.1
Gene
ATP5J2-PTCD1
Gene Name
ATP5J2-PTCD1 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001198879.1 Intron NP_001185808.1
Gene
CPSF4
Gene Name
cleavage and polyadenylation specific factor 4
There are no transcripts associated with this gene.

View Full Product Details