Product Details

SNP ID
rs6978652
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:64804946 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCCGTGTTTCTCATGTCGAGAGTAG[C/T]GGTGCACTTTGGGTGTTTAAAAAAA
Phenotype
MIM: 604080
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF138 PubMed Links

Gene Details

Gene
ZNF138
Gene Name
zinc finger protein 138
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001160183.2 Intron NP_001153655.1
NM_001271637.1 Intron NP_001258566.1
NM_001271638.1 Intron NP_001258567.1
NM_001271639.1 Intron NP_001258568.1
NM_001271640.1 Intron NP_001258569.1
NM_001271649.1 Intron NP_001258578.1
NM_006524.3 Intron NP_006515.2

View Full Product Details