Product Details
- SNP ID
-
rs558660
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:59836421 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- AGAGTTTGAGGAATCCATACCAAAC[A/G]CTAGTAATCTCTGGAGTAATAAGTT
- Phenotype
-
MIM: 609342
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
GIF
PubMed Links
Gene Details
- Gene
- GIF
- Gene Name
- gastric intrinsic factor
View Full Product Details