Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001202504.1 | 13 | UTR 5 | NP_001189433.1 | ||
NM_001318511.1 | 13 | Intron | NP_001305440.1 | ||
NM_004715.4 | 13 | Intron | NP_004706.3 | ||
NM_048368.3 | 13 | Intron | NP_430255.2 | ||
XM_011526261.1 | 13 | Intron | XP_011524563.1 | ||
XM_017026078.1 | 13 | Intron | XP_016881567.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
XM_011526290.2 | 13 | UTR 5 | XP_011524592.1 | ||
XM_011526292.2 | 13 | Intron | XP_011524594.1 | ||
XM_017026112.1 | 13 | UTR 5 | XP_016881601.1 |